Learn More
Motic™ Wafer Holder, BA310MET-T
Specifically designed to hold wafers for detailed inspection. Motic™ Wafer Holder, BA310-Met-T is an optional stage clip accessory.
Brand:  Motic™ 1101001904211
Product Code. 15435262
  • 2885.00 NOK / Each
Estimated Shipment: 24-03-2025
to see stock.
Explore more special offers
Description
  • Microscope stage accessory.
  • Stroke: 6in. X 4in.
Specifications
Specifications
Wafer Holder
Stroke: 6in. x 4in.