missing translation for 'onlineSavingsMsg'
Learn More
VU6012962, MedChemExpress Product Code.: 16444828

VU6012962, MedChemExpress

Product Code. 16444828
10 mg, 10mg
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16444828

Brand: Medchem Express HY114403

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress VU6012962 is an orally bioavailable and CNS-penetrant metabotropic glutamate receptor 7 negative allosteric modulator (mGlu7 NAM) with an IC50 of 347 nM.

VU6012962 is an orally bioavailable and CNS-penetrant metabotropic glutamate receptor 7 negative allosteric modulator (mGlu7 NAM) with an IC50 of 347 nM.

Specifications

Chemical Name or Material VU6012962
CAS 2313526-86-0
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C21H19F3N4O4
Quantity 10 mg
Solubility Information DMSO : 125 mg/mL (278.77 mM; Need ultrasonic)
SMILES O=C(C1=CC=C(OCC2CC2)C(OC)=C1)NC3=C(N4C=NC=N4)C=CC(OC(F)(F)F)=C3
Molecular Weight (g/mol) 448.4
Formula Weight 448.4
Percent Purity 99.89%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
VU6012962, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.