Learn More
VPS39 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 16219-1-AP-150UL
 View more versions of this product
Product Code. 16845774
  • 4796.50 NOK / 150µL
Explore more special offers
Description
Description
VPS39 is the mammalian homolog of the yeast vacuolar protein VAM6, that in mammals, promotes lysosome clustering and fusion in vivo. VPS39 is also thought to modulate the TGF-beta response by coupling the TGF-beta receptor complex to the Smad pathway. Similar to the related protein TRAP1, the VPS39 protein is essential for early embryonic development, with mice lacking VPS39 dying at or before E6.5.
Specifications
Specifications
VPS39
Polyclonal
Unconjugated
VPS39
hVam6p, KIAA0770, TLP, VAM6, Vam6/Vps39 like protein, VPS39
Rabbit
Antigen Affinity Chromatography
RUO
23339, 269338
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation
0.14 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8R5L3, Q96JC1
VPS39
VPS39 Fusion Protein Ag9267
150 μL
Primary
Human, Mouse