Learn More
Scientific Industries SI™ TurboMix Attachment
Provides extremely vigorous mixing through a combination of vortexing and impact collision. Scientific Industries SI™ TurboMix Attachment is ideal for aggressive and rapid dissolution, resuspension or mixing of samples
Brand: Scientific Industries SI™ SI-0564
This item is not returnable.
View return policy
Description
- Permanently mounts to most existing Vortex-Genie 2 family of mixers and the Vortex-Genie Pulse.
- Holds up to twelve 1.5mL or 2.0mL microtubes
- Ideal for aggressive and rapid dissolution, resuspension or mixing of samples
- Convenient and effective for cell disruption in glass bead procedures.
Specifications
TurboMix Attachment | |
Most Vortex-Genie 2 family of mixers, Vortex-Genie Pulse | |
12 |
Product Content Correction
Product Title
Scientific Industries SI™ TurboMix Attachment > Holds: 12 x 1.5 to 2mL Screw Top or Snap Top Tubes