Learn More
VACUUBRAND™ VMS-B Module For Vacuum Controller CVC 3000 to Switch a Vacuum Pump
The Vacuum-Management-System module VMS-B switches a vacuum pump according to actual demand from one or two applications;It is operated by one or two vacuum controllers CVC 3000. If two CVC 3000 are connected to the VMS-B it switches off the pump only if both applications do not need a vacuum supply anymore.
Brand:  VACUUBRAND™ 20676030
Product Code. 11990372
  • 5035.00 NOK / Each
Estimated Shipment: 08-05-2025
to see stock.
Explore more special offers
Add to basket
Specifications
For Vacuum Controller CVC 3000 to Switch a Vacuum Pump
1 Each
VMS-B Module
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
VACUUBRAND™ VMS-B Module For Vacuum Controller CVC 3000 to Switch a Vacuum Pump > For use with: For Vacuum Controller CVC 3000 to Switch a Vacuum Pump