Learn More
Sartorius Vivaspin™ Turbo 15 RC Ultrafiltration Centrifugal Concentrators
Vivaspin™ Turbo 15 Regenerated Cellulose (RC) ultrafiltration centrifugal concentrators for protein, DNA, nanoparticle and virus concentration for 4-15 mL sample volumes. Acquire the fastest sample concentration with the highest recoveries for your protein samples.
2750.00 NOK - 10030.00 NOK
Specifications
Disposable | Single-use |
---|---|
Filter Type | Ultrafiltration |
For Use With (Equipment) | Centrifuge |
Material (Membrane) | Regenerated Cellulose |
Sample Volume | 15 mL |
Product Code | Brand | MWCO | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | MWCO | Quantity | Price | Quantity & Availability | |||||
16646942
![]() |
Sartorius
VS15TR11 |
5 kDa | 12/Pk. |
2750.00 NOK
Pack of 12 |
Estimated Shipment: 11-07-2025
Log in to see stock. |
|||||
16656942
![]() |
Sartorius
VS15TR12 |
5 kDa | 48/Pk. |
10030.00 NOK
Pack of 48 |
Estimated Shipment: 25-04-2025
Log in to see stock. |
|||||
16626942
![]() |
Sartorius
VS15TR01 |
10 kDa | 12/Pk. |
2750.00 NOK
Pack of 12 |
Estimated Shipment: 11-06-2025
Log in to see stock. |
|||||
16636942
![]() |
Sartorius
VS15TR02 |
10 kDa | 48/Pk. |
10030.00 NOK
Pack of 48 |
Estimated Shipment: 11-06-2025
Log in to see stock. |
|||||
16666942
![]() |
Sartorius
VS15TR21 |
30 kDa | 12/Pk. |
2750.00 NOK
Pack of 12 |
Estimated Shipment: 02-07-2025
Log in to see stock. |
|||||
16676942
![]() |
Sartorius
VS15TR22 |
30 kDa | 48/Pk. |
10030.00 NOK
Pack of 48 |
Estimated Shipment: 11-07-2025
Log in to see stock. |
|||||
16686942
![]() |
Sartorius
VS15TR31 |
50 kDa | 12/Pk. |
2750.00 NOK
Pack of 12 |
Estimated Shipment: 11-06-2025
Log in to see stock. |
|||||
16696942
![]() |
Sartorius
VS15TR32 |
50 kDa | 48/Pk. |
10030.00 NOK
Pack of 48 |
Estimated Shipment: 11-06-2025
Log in to see stock. |
|||||
16606952
![]() |
Sartorius
VS15TR41 |
100 kDa | 12/Pk. |
2750.00 NOK
Pack of 12 |
Estimated Shipment: 11-07-2025
Log in to see stock. |
|||||
16616952
![]() |
Sartorius
VS15TR42 |
100 kDa | 48/Pk. |
10030.00 NOK
Pack of 48 |
Estimated Shipment: 02-07-2025
Log in to see stock. |
|||||
Description
Vivaspin™ Turbo centrifugal concentrators offer the optimal choice for maximum concentration or buffer exchange applications, with their broad range of MWCOs and market unique membrane choice of RC and PES membrane.
The Vivaspin™ Turbo 15 RC concentrators are the Regenerated Cellulose concentrators with the highest flow rates on the market due to their large, twin vertical membranes and sleek internal profile, which minimize protein polarization and subsequent fouling of the membrane and ensures maximum process speeds right down to the last 100 μL.
- Unique optimal membrane choice (PES and RC)
- Complete recoveries
- Fastest concentrations
- Highest chemical resistance
- Easy retentate collection
- Graduated and markable
Specifications
Single-use | |
Centrifuge | |
15 mL | |
Polypropylene | |
15 mL |
Ultrafiltration | |
Regenerated Cellulose | |
Non-sterile | |
Centrifugal Concentrator | |
77 mm |