Learn More
Sartorius Vivapore 10
Vivapore 10 solvent absorption concentrators are disposable ultrafilters for macromolecule concentration from up to 10 mL aqueous samples.
Brand: Sartorius VP-S010P0008-3
2491.84 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Concentrate equipment-free
- Fill and go
- Tailored process monitoring
- Maintained sample integrity
- Sensitive molecule friendly
Specifications
Ultrafiltration | |
PES | |
10 mL | |
Solvent Absorption Concentrator |
SAN | |
7.5 kDa | |
Non-sterile | |
10 mL |