missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vitlab™ VITsafe™ Wide-mouth Safety Wash Bottles
Click to view available options
Capacity (Metric):
250 mL
500 mL
Chemical Label:
Ethyl Acetate
Pentane
Description
- Bottles made from PE-LD (or PP for Acetone and MEK), spray tube from PP.
- With colored VENT-CAP which supports easy visual identification of the media.
- Chemical formula, CAS No., hazard pictogram, signal word
- Risk phrases (H phrases), safety phrases (P phrases), as well as NFPA Code
Specifications
Specifications
Capacity (Metric) | 250 mL |
Material | LDPE |
Thread Size (Metric) | GL 45 |
Closure Color | Red |
Height (Metric) | 146 mm |
Chemical Label | Ethyl Acetate |
Diameter (Metric) | 58 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Vitlab™ VITsafe™ Wide-mouth Safety Wash Bottles >
Spot an opportunity for improvement?Share a Content Correction