Learn More
Ismatec™ Viton™ Microbore 2-Stop Tube Sets
Choose the size and formulation to suit your flow rate and application needs.
Brand:  Ismatec™ 96428-48
 View more versions of this product
Product Code. 12688093
  • 4060.00 NOK / Pack of 12
Estimated Shipment: 28-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Genuine Ismatec™ tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
Specifications
Specifications
FDA approved; Prop 65 NA (no Prop 65 chemicals)
2.79 mm
1.25 ft.
Pump Series L/S
12 / Pk.
1/8 in.
150°C
Stopped Pump Tubing
Viton
0.38 m
0°C
200 mL/min
25.3 psi, 1.75 bar