missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VistaLab™ Technologies MLA™ Brand Precision Pipetters

Ten fixed-volume models from 10 to 1000μL
Brand: VistaLab™ Technologies 1112
This item is not returnable.
View return policy
Description
- 'To deliver' capability ensures volume accuracy over a wide range of viscosities
- Ideal for repetitive delivery of reagents when high precision and accuracy are required
- Sleeves are color coded for easy volume identification
- Capacity engraved on bonnet
Order Info
Additional sizes available. Contact your Fisher Scientific Customer Service Representative for more information.
Specifications
±1.00% | |
Silver | |
Micropipette Tip | |
F1 | |
Non-sterile | |
150 μL | |
Limited Lifetime | |
2 in. |
Not Autoclavable | |
Reusable | |
Aluminum, Stainless Steel | |
1 | |
Precision Pipette | |
Nozzle Insert, Calibration Key | |
9 in. | |
1.5 in. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
VistaLab™ Technologies MLA™ Brand Precision Pipetters > Volume: 150μL; Color Code: Silver
Spot an opportunity for improvement?Share a Content Correction