missing translation for 'onlineSavingsMsg'
Learn More

VistaLab™ Technologies MLA™ Brand Precision Pipetters
GREENER_CHOICE

Ten fixed-volume models from 10 to 1000μL

Brand:  VistaLab™ Technologies 1027

 View more versions of this product

Product Code. 11905792

  • 3140.00 NOK / Each
Estimated Shipment: 03-07-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • 'To deliver' capability ensures volume accuracy over a wide range of viscosities
  • Ideal for repetitive delivery of reagents when high precision and accuracy are required
  • Sleeves are color coded for easy volume identification
  • Capacity engraved on bonnet

Order Info

Additional sizes available. Contact your Fisher Scientific Customer Service Representative for more information.

Specifications

Specifications

±1%
Purple
Micropipette Tip
F1
Non-sterile
250 μL
1 Ea.
9 in.
1.5 in.
Not Autoclavable
Reusable
Aluminum, Stainless Steel
1
Precision Pipette
Nozzle Insert, Calibration Key
Limited Lifetime
2 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
VistaLab™ Technologies MLA™ Brand Precision Pipetters > Volume: 250μL; Color Code: Purple

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.