Learn More
SCHOTT AG Lighting and Imaging™ VisiLED Controller
The VisiLED Controllers are the core of the VisiLED system to control the VisiLED light heads.
5610.00 NOK
Spécification
For Use With (Application) | Stereo Microscopy Illumination |
---|---|
For Use With (Equipment) | VisiLED Ringlights or Transmitted Light Stage |
Code produit | Marque | Type | For Use With (Equipment) | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Type | For Use With (Equipment) | Prix | Quantité et disponibilité | |||||
16637105
|
SCHOTT AG Lighting and Imaging™
400080 |
VisiLED Standard Controller MC1100 | VisiLED Ringlights or Transmitted Light Stage |
5610.00 NOK
Each |
Estimated Shipment: 08-05-2025
Log in to see stock. |
|||||
Description
- The compact MC1100 allows the settings of various illumination parameters including light intensity, segment modes and change of illumination direction while the MC1500 offers this for two light heads.
- Extra-fine smooth dimming.
- MC1500 offers an external trigger to start strobe function frequencies netween 15-5 KHz.
- MC1500 offers optional PC control via RS232 and footswitch control.
- Advanced setting options for optimum contrast.
- Ergonomics and blindfold operation.
- MC1500 offers control of two channels.
- Save up to four complex illumination scenarios with MC1500.
Spécification
Stereo Microscopy Illumination | |
VisiLED Ringlights or Transmitted Light Stage |
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
SCHOTT AG Lighting and Imaging™ VisiLED Controller