Learn More
Brady™ Vinyl Cloth Labels for M611, BMP61 and BMP71
Once applied, this repositionable vinyl cloth label is there to stay...unless you want to change it.
570.00 NOK - 1080.00 NOK
Specifications
Autoclavable | Not Autoclavable |
---|---|
Adhesive Type | Repositionable Rubber |
Color | White |
Finish | Semi-Gloss |
Material | Vinyl Cloth (B-498) |
Product Code | Brand | Height (Metric) | Shape | Vertical Repeat (Metric) | Width (Metric) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Height (Metric) | Shape | Vertical Repeat (Metric) | Width (Metric) | Price | Quantity & Availability | ||
11552975
|
Brady™
062668 |
6.4 mm | Rectangle | 9.5 mm | 38.1 mm | N/A | |||
11582965
|
Brady™
062661 |
7 mm | Rectangle | 10.2 mm | 25.4 mm | N/A | |||
11552965
|
Brady™
062657 |
12.7 mm | Square | 15.9 mm | 12.7 mm | N/A | |||
12343478
|
Brady™
062659 |
19.1 mm | Rectangle | 9.5 mm | 6.4 mm | N/A | |||
11512975
|
Brady™
062664 |
25.4 mm | Rectangle | 22.2 mm | 19.1 mm | N/A | |||
11522975
|
Brady™
062665 |
25.4 mm | Square | 28.6 mm | 25.4 mm | N/A | |||
11572975
|
Brady™
062670 |
38.1 mm | Rectangle | 22.2 mm | 19.1 mm | N/A | |||
11542975
|
Brady™
062667 |
101.6 mm | Rectangle | 104.8 mm | 25.4 mm | N/A | |||
11512985
|
Brady™
062674 |
152.4 mm | Rectangle | 155.6 mm | 38.1 mm | N/A | |||
Description
- Adhesive and cloth backing offers excellent holding power, while allowing for clean removal and repositioning
- Sticks to any clean, dry surface
- Specially formulated topcoat for excellent print quality
- Semi-gloss finish for crisp, clean look
Specifications
Not Autoclavable | |
White | |
Vinyl Cloth (B-498) | |
Non-sterile | |
BMP™61, BMP™71, BradyPrinter M611, TLS 2200™ and TLS-PC LINKTM Thermal Labeling Systems | |
-40°C to +80°C |
Repositionable Rubber | |
Semi-Gloss | |
Thermal Transfer | |
Barcode Labeling, Circuit Board Labeling, Equipment Labeling, Wire and cable Labeling | |
1 | |
0.185 mm |