missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vinculin Antibody (VCL/8533R), Alexa Fluor™ 350, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21067AF350
This item is not returnable.
View return policy
Description
Vinculin Monoclonal antibody specifically detects Vinculin in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
Specifications
Vinculin | |
Monoclonal | |
Alexa Fluor 350 | |
CMD1W, CMH15, Metavinculin, MVCL, vinculin | |
Recombinant fragment (around aa 174-322) of human Vinculin protein (exact sequence is proprietary) | |
0.1 mL | |
Cell Biology, Cellular Markers, Phospho Specific | |
7414 | |
Store at 4°C in the dark. | |
IgG κ |
Western Blot, Immunohistochemistry (Paraffin) | |
VCL/8533R | |
50 mM Sodium Borate | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 6
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Vinculin Antibody (VCL/8533R), Alexa Fluor™ 350, Novus Biologicals™ > 0.1 mL, Alexa Fluor 350
Spot an opportunity for improvement?Share a Content Correction