Learn More
Vileda Professional™ Telescopic Handle, short
For use in controlled environments for effective cleaning. Vileda Professional Telescopic Handle, short with adjustable length of 50 – 90cm.
Brand:  Vileda Professional™ 111384
Product Code. 17802486
  • 3050.00 NOK / Pack of 10
Estimated Shipment: 15-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Constructed of aluminum
  • Adjustable length of 50 - 90cm
  • Extends with twist of the wrist
  • Anti-slip top grip to reduce slipping on walls
  • Ergonomic grip allows blood circulation in hands and fingers, thereby reducing fatigue
  • Resists autoclave, gamma and ETO processes
  • Fits CE Squeegee and other commonly used frames
Specifications
Specifications
Telescopic Handle, Short
Blue, Silver
Swep Duo Plus Frame
Floors, Walls and Ceilings, ISO 5, LAR, Production, Labs