Learn More
Fisherbrand™ Vial Decapper Pliers
Fisherbrand™ Vial Decapper Pliers are designed to easily remove aluminum crimp tops from vials and bottles.
889.00 NOK - 962.00 NOK
Specifications
Material | Stainless Steel |
---|---|
Certifications/Compliance | ISO 9002, cGMP |
Type | Plier-style Decapper |
Product Code | Brand | Diameter (Metric) Outer | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outer | Price | Quantity & Availability | ||||||
17837658
|
Fisherbrand™
13-820-043 |
8 mm |
962.00 NOK
Each |
Estimated Shipment: 02-09-2025
Log in to see stock. |
||||||
16153167
|
Fisherbrand™
13820047 |
30 mm |
889.00 NOK
Each |
Estimated Shipment: 16-07-2025
Log in to see stock. |
||||||
Description
Fisherbrand™ Vial Decapper Pliers are designed to easily remove aluminum crimp tops from vials and bottles. Available in single- or double-action formats, all pliers are smooth and simple to operate. Durable, high-quality construction ensures consistent, dependable performance.
- Easily remove aluminum crimp-tops from vials and bottles
- Smooth and simple to operate
- Durable, high-quality construction offering consistent, dependable performance
- Padded-handles allow for maximum grip and comfort to relieve stress tension from hands
- Made from one-piece, corrosion-resistant ASTM-standard stainless steel
- Compatible with Gamma/ETO/Autoclavable sterilization methods
- Overall length: 159 mm (6.25 in.)
Certifications
ISO 9001-02, GMP, cGMP
Specifications
Stainless Steel | |
ISO 9002, cGMP | |
Plier-style Decapper |