Learn More
Tocris Bioscience™ Verteporfin
YAP inhibitor
Brand:  Tocris Bioscience™ 5305/10
Product Code. 18779967
  • 2740.00 NOK / 10mg
Explore more special offers
Description
Description
Verteporfin is a YAP inhibitor; disrupts YAP-TEAD interactions. Enhances trypsin cleavage of YAP (EC50 = 100 nM). Inhibits growth and proliferation of retinoblastoma cells. Also suppresses cancer stem cell properties in vitro. Suppresses YAP-induced liver overgrowth in mice.
Specifications
Specifications
95%
10 mg
718.79
Visudyne
Verteporfin
Verteporfin
For Research Use Only