Learn More
Tocris Bioscience™ Verteporfin
YAP inhibitor
Brand: Tocris Bioscience™ 5305/10
This item is not returnable.
View return policy
Description
Verteporfin is a YAP inhibitor; disrupts YAP-TEAD interactions. Enhances trypsin cleavage of YAP (EC50 = 100 nM). Inhibits growth and proliferation of retinoblastoma cells. Also suppresses cancer stem cell properties in vitro. Suppresses YAP-induced liver overgrowth in mice.Specifications
95% | |
10 mg | |
718.79 |
Visudyne | |
Verteporfin | |
Verteporfin |