Learn More
Hirschmann™ Versilon™ 2001 Multichannel Stopper Tubes
For use in peristaltic pumps with multichannel pump heads.
Brand:  Hirschmann™ 95214725
Product Code. 30090411
  • 3015.00 NOK / Pack of 6
Only null left
Estimated Shipment: 18-08-2025
Description
  • Always comes with an RFID-chip carrier on which important tubing parameters (inner diameter, wall thickness, service life expectancy) are saved and available for the pump’s software
  • Pump is capable to start instantly in the flow rate and target volume mode through the available tubing works-factor with this RFID-chip carrier and the pump head identification
  • Stopper 104
  • Violet-white in color
Specifications
Specifications
381 mm
ID: 2.79 mm; Stopper 104
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Hirschmann™ Versilon™ 2001 Multichannel Stopper Tubes > ID: 2.79 mm