Learn More
3M™ Versaflo™ Powered Air Turbo Unit, TR-602E
Click to view available options
:
Air Turbo Unit; Battery Type: Lithium-Ion
Description
- Combines with 3M™ Versaflo™ components to build a complete system
- Three selectable flow rates (185 L/min., 205 L/min. or 225 L/min.) for maximum comfort with a steady airflow
- Visual, vibratory and audible warning alarms show you low battery and low air flow
- Battery charge, airflow and particulate filter loading status indicators
- Choice of battery types: standard or high capacity
Specifications
Specifications
Battery Type | Lithium-Ion |
Color | Light Gray |
Product Type | Powered Air Turbo Unit |
Quantity | 1 Ea. |
Product Content Correction
Product Title
3M™ Versaflo™ Powered Air Turbo Unit, TR-602E >