Description
- Flame resistant, heat resistant, and highly durability breathing tube cover
- Quick release swivel enables one-handed connection and release
- Helps lengthens the life of the tube and protects it from flying sparks
- Suitable for use with 3M™ Breathing Tubes, BT-Series
Specifications
Specifications
Certifications/Compliance | CE Approved |
Color | Black |
Material | Oxidized Polyacrylonitrile |
Product Type | Breathing Tube Cover |
For Use With (Equipment) | 3M™ BT-Series Breathing Tubes |
Product Content Correction
Product Title
3M™ Versaflo™ Breathing Tube Cover >