Learn More
Linden™ Vernier Callipers with Micrometer Screw Gauge
Can be used for internal, external and depth measurement. Linden™ Vernier Callipers with Micrometer Screw Gauges have a micrometer screw gauge supplied in case.
Brand:  Linden™ H.DR39493
 View more versions of this product
Product Code. 13015903
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • General purpose with thumb wheel for fine adjustment
  • Metric and imperial dual graduations
  • Available in a range of lengths
Specifications
Specifications
115 mm
Documents
One moment while we fetch your results.
Product Title
Linden™ Vernier Callipers with Micrometer Screw Gauge > Length: 115mm