missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ VER 246608

Potent and selective pyruvate dehydrogenase kinase (PDK) 1/2/3/4 inhibitor

2290.00 NOK - 9025.00 NOK

Specifications

Chemical Name or Material N-[4-(2-Chloro-5-methyl-4-pyrimidinyl)phenyl]-N-[[4-[[(2,2-difluoroacetyl)amino]methyl]phenyl]methyl]-2,4-dihydroxybenzamide
CAS 1684386-71-7
Target Other Kinase Inhibitors
Molecular Formula C28H23ClF2N4O4
Purity 0.98
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18778976
View Documents
Tocris Bioscience™
6604/10
10 mg
2290.00 NOK
10mg
Please to purchase this item. Need a web account? Register with us today!
18759926
View Documents
Tocris Bioscience™
6604/50
50 mg
9025.00 NOK
50mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

VER 246608 is a potent and ATP-competitive inhibitor of pyruvate dehydrogenase kinase (PDK; IC50s = 35, 40, 84, and 91 nM for PDK-1, PDK-3, PDK-2, and PDK-4 respectively). It is selective for PDKs over a panel of 97 kinases and HSP90. VER 246608 increases pyruvate dehydrogenase complex activity, oxygen consumption and reduces glycolytic activity; it reduces the production of L-lactate in PC3 cells in D-glucose and L-glutamine depleted media. In PC3 cells, VER 246608 induces cell cycle arrest at the G1 phase under glucose or serum-depleted conditions characteristic of the tumor microenvironment.
Specifications

Specifications

N-[4-(2-Chloro-5-methyl-4-pyrimidinyl)phenyl]-N-[[4-[[(2,2-difluoroacetyl)amino]methyl]phenyl]methyl]-2,4-dihydroxybenzamide
Other Kinase Inhibitors
0.98
1684386-71-7
C28H23ClF2N4O4
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tocris Bioscience™ VER 246608

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.