missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VEGFA Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1415.13 NOK - 3828.53 NOK
Specifications
Antigen | VEGFA |
---|---|
Concentration | 233 μg/mL |
Applications | Western Blot, Immunohistochemistry, Flow Cytometry, Co-immunoprecipitation (Co-IP), ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16844865
|
Proteintech
26381-1-AP-20UL |
20 μL |
1415.13 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16834865
|
Proteintech
26381-1-AP-150UL |
150 μL |
3828.53 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The VEGFA antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human VEGFA. This antibody recognizes human antigen. The VEGFA antibody has been validated for the following applications: FC, IHC, WB, ELISA analysis.
Specifications
VEGFA | |
Western Blot, Immunohistochemistry, Flow Cytometry, Co-immunoprecipitation (Co-IP), ELISA | |
Unconjugated | |
Human | |
IgG |
233 μg/mL | |
Polyclonal | |
Rabbit | |
7422 | |
Primary |