Learn More
Novus Biologicals™ VECTASTAIN(R) ABC anti-Rabbit IgG AP Immunodetection Kit
Immunodetection Kit
Brand:  Novus Biologicals™ AK-5001-NB
Product Code. 18354138
  • 6355.00 NOK / 1 set
Explore more special offers
Specifications
Store at 4°C. Do not freeze.
Western Blot, ELISA, Immunohistochemistry (IHC), Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Frozen), ISH, ELISpot
Rabbit
1 Kit
Product Title
Novus Biologicals™ VECTASTAIN(R) ABC anti-Rabbit IgG AP Immunodetection Kit > 1 Kit