Learn More
Cole-Parmer™ VapLock™ Solvent Waste Manifold Caps for Bottles and Carboys
VapLock™ Solvent Waste Manifold Caps for Bottles and Carboys connect to waste containers for a secure closed system
Brand: Cole-Parmer™ 12018-84
This item is not returnable.
View return policy
Description
- Select from a variety of cap sizes to fit your container
- Different materials, numbers and sizes of ports help customize to your application
Warranty
None
Specifications
Solvent Safety | |
PTFE | |
1/2 in. NPT(F), 1/4 in. NPT(F), 1/4 in.-28 UNF(F) |
Solvent-Compatible Containers | |
DIN 45 | |
Solvent Waste Manifold, PTFE, DIN45, 1 x 1/2 inNPTF, 3 x 1/4 inNPTF, 6 x 1/4 in.-28 UNFF; 1/ea |
Product Content Correction
Product Title
Cole-Parmer™ VapLock™ Solvent Waste Manifold Caps for Bottles and Carboys > PTFE, DIN45, 1x1/2 inNPTF, 3x1/4 inNPTF, 6x1/4 in-28UNFF; 1/ea