missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vanadium Sulfate (Vanadyl), MP Biomedicals
Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 g
Specifications
Specifications
CAS | 27774-13-6 |
Quantity | 100 g |
Molecular Formula | O5SV |
Synonym | vanadic sulfate, vanadium oxide sulphate, oxovanadium 2+ sulfate, vanadium oxysulfate, oxovanadium iv sulfate, voso4, vanadin iv oxide sulfate, vanadium, oxosulfato, vanadium, oxysulfato, vanadyl sulfate vso5 |
InChI Key | UUUGYDOQQLOJQA-UHFFFAOYSA-L |
SMILES | [O-]S(=O)(=O)[O-].O=[V+2] |
IUPAC Name | oxovanadium(2+);sulfate |
Molecular Weight (g/mol) | 162.996 |
PubChem CID | 34007 |
ChEBI | CHEBI:33146 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Vanadium Sulfate (Vanadyl), MP Biomedicals >
Spot an opportunity for improvement?Share a Content Correction