missing translation for 'onlineSavingsMsg'
Learn More

Vanadium AAS Standard, 1000 mg/L V in H2SO4 2%, Honeywell Fluka™

409.34 NOK - 668.12 NOK

Specifications

Chemical Name or Material Vanadium AAS Standard
Density 1.02 g/cm3
pH <2
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16694404
View Documents
Honeywell Fluka
E4081-100ML
100 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16604414
View Documents
Honeywell Fluka
E4081-500ML
500 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Honeywell Fluka™ analytical standards offer a growing range of high quality inorganic and organic reference materials for the European market produced according to ISO 9001. The products are suitable for a wide range of industries and applications and come complete with a detailed Certificate of Analysis (CoA) in every delivery, and are eligible for free expert technical support ensuring you have the information you need to quickly start your analysis and pass an audit.

Specifications

Specifications

Vanadium AAS Standard
1.02 g/cm3
<2
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 760

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Vanadium AAS Standard, 1000 mg/L V in H2SO4 2%, Honeywell Fluka™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.