Learn More
VACUUBRAND™ Valve and Diaphragm Set
Brand: VACUUBRAND™ 20696834
2351.69 NOK valid until 2025-04-13
Use promo code "24122" to get your promotional price.
Description
Specifications
For MV 2, MD4 Vario | |
1 Each | |
Valve and Diaphragm Set |
Product Suggestions
Customers who viewed this item also viewed
Special Offers
-
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13
New Offer Save Up to 92%