Learn More
OHAUS™ Valor™ 2000 Scale
OHAUS™ Valor™ 2000 Scale offers large LED front and rear displays, making it ideal for use in dry or washdown environments.
Brand: OHAUS™ 30072315
3626.62 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- The sturdy polycarbonate keypad offers protection from knives and other sharp instruments used in food preparation, processing and portioning applications.
- A unique IP68 flow-thru design ensures that the NSF-certified Valor 2000 will drain moisture in the scale when used in wet conditions.
- Stabilization time is less than a half-second.
- Large front and rear LED displays allow for easy viewing so two operators can share one scale.
Specifications
50 Hours with Rechargeable Battery | |
6.00 kg | |
1.000000 g | |
V22PWE6T | |
1.000000 g | |
-10°C to +40°C | |
256 x 280 x 121 mm |
ABS housing with stainless steel platform (PW), stainless steel housing and platform (XW), up-front level indicator | |
Dual LED | |
1.000000 g | |
IPX8 | |
0.500 sec. | |
Gram, Kilogram, Ounce, Pound, Pound:Ounce | |
5.000 kg |
Product Content Correction
Product Title
OHAUS™ Valor™ 2000 Scale > Model: V22PWE6T