Learn More
BINDER™ Pump Chamber with VAP 2 Vacuum Pump
Pump Chamber with VAP 2 Vacuum Pump - 230 V / 50-60 Hz option model, pump chamber for vacuum pumps, with switchable power socket. Chemical membrane pump with a nominal air flow of 4.0 m³/hour, final pressure 1.8 mbar
Brand:  BINDER™ 8012-1965
83426.43 NOK valid until 2024-12-31
Use promo code "21853" to get your promotional price.
 View more versions of this product
Product Code. 16664392
  • 89900.00 NOK / Each
Estimated Shipment: 14-03-2025
to see stock.
Explore more special offers
Description
Pump chamber:
  • Made of powder-coated sheet metal, with large viewing window
Pump:
  • Nominal air flow: 4.0 m³/hr.
  • End vacuum: 1.8 mbar
  • Pump membrane made of highly resistant PEEK plastic
  • Noise level < 50 dB(A)
Specifications
Specifications
1011.45 mbar below atmospheric pressure
Pump chamber with VAP 2 vacuum pump - 230 V / 50-60 Hz option model, Pump chamber for vacuum pumps, with switchable power socket. Chemical membrane pump with a nominal air flow of 4.0 m3/hour, final pressure 1,8 Mbar
50/60 Hz
IP55
BINDER Chambers VD 56
230 V, 50/60 Hz
4.0 m3/hr.
Vacuum Pump
2 Mbar
230 V