Learn More
VACUUBRAND™ VACUU·SELECT Vacuum Controller
The application-based VACUU·SELECT Controller with a touch screen display is easy-to-use, versatile, and efficient for all common vacuum processes in chemistry labs. Create unique applications with simple drag-and-drop editing, and detect boiling points for solvent evaporation.
Brand: VACUUBRAND™ 20700000
19097.31 NOK valid until 2024-12-31
Use promo code "21805" to get your promotional price.
This item is not returnable.
View return policy
Description
- Graphic user interface simplifies laboratory work
- Predefined applications save time and guarantee reproducible results
- Application editor allows easy customization
- Integrated help function
- Manual set-point-control for simple processes
- Touch screen display can be operated even when wearing safety gloves
- Ceramic vacuum sensor
- Measuring Range: 0.1 to 1080 mbar (0.1 to 810) torr)
- Measurement Principle: Ceramic Diaphragm (Alumina), Capacitive, Gas Independant, Absolute Pressure
- Choose from 14 languages
Specifications
± 1 Mbar/hPa/torr | |
Touchscreen | |
Ceramic Vacuum |
VACUU·SELECT | |
1 Ea. | |
Vacuum Controller |