Learn More
Integra Biosciencesā„¢ VACUSAFEā„¢ Lid
Specifically designed lid for the VACUSAFEā„¢ aspiration system. Integra Biosciencesā„¢ VACUSAFEā„¢ Lid includes integrated tubing connections.
Brand:  Integra Biosciencesā„¢ 158372
 View more versions of this product
Product Code. 10098601
  • 1673.70 NOK / Each
Only null left
Estimated Shipment: 31-07-2025
Description
  • Suitable for the VACUSAFEā„¢ vacuum aspiration system
  • Integrated tubing connections
  • Quick couplings and level detection in some models
Specifications
Specifications
4 L VACUSAFE Aspiration Bottle
Tubing Fittings
Lid
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Integra Biosciencesā„¢ VACUSAFEā„¢ Lid > For 4L VACUSAFEā„¢ bottle