Learn More
Cytiva VacuCap™ 90 Vacuum Filtration Devices
Innovative bottle-top filters for fast vacuum filtration of 100 mL to 5 L of aqueous solutions.
Brand: Cytiva 4624
2335.97 NOK valid until 2025-07-04
Use promo code "24147" to get your promotional price.
Description
- This product is not approved for use in medical, clinical, surgical or other patient applications
Specifications
Supor (Hydrophilic PES) | |
Sterile | |
90 mm |
0.45 μm | |
60 cm2 |
Promotions
-
Multiple Manufacturers
Quality Analysis and Quality Control Testing
Save up to 59% on some of our most popular productsPromo Code: 24140Expires: 2025-08-24
New Offer Save Up to 59%