Learn More
uvex i-3 Safety Goggles
Click to view available options
Frame Color:
Anthracite Blue
Black, Light Gray
Lens Tint:
Clear
Gray
Description
Specifications
Specifications
Certifications/Compliance | EN 170 |
Lens Type | Uvex™ supravision excellence |
Lens Coating | Anti-fog |
Description | Shield-lens spectacle |
Lens Tint | Gray |
Product Type | Ergonomic Safety Glasses |
Frame Color | Black, Light Gray |
Material | Polycarbonate |
Disposable | Reusable |
Lens Material | Polycarbonate |