Learn More
Eppendorf™ UVette™ Starter Set
Individually packaged UVettes™ are DNA, RNA and protein free, and are protected from scratches and dust
Brand:  Eppendorf™ 4099100007
Additional Details : Weight : 0.31000kg
Product Code. 10081992
  • 1115.00 NOK / 1 set
Estimated Shipment: 29-07-2025
Description
Individually packaged UVettes™ are DNA, RNA and protein free, and are protected from scratches and dust.
  • Disposable cuvette for UV light measurements
  • Fully transparent plastic for all VIS applications
  • UV and VIS transparent between 220nm to 1, 600nm
  • Suitable for measuring small volumes (≥50μL)
  • Individually blister-packed for contamination free work processes
  • Volume markings for 500μL and 1,000μL
  • Optimal filling guaranteed by funnel shaped tube base
  • Frosted grip allows for labelling of the cuvette
  • Deep-lying optical window prevents scratches
  • Samples with a normal concentration can be measured using the 10mm optical path length, for samples with a higher concentration, simply rotate the UVette™ by 90°C and use the shorter 2mm optical path length
  • Can be used as is with the BioPhotometer, and with all other spectrophotometers with the aid of an adapter
  • Adapter aperture: 1.5mm x 1.5mm (h x w)
Specifications
Specifications
Starter Set
For measurements in the UV/Vis range with transparency from 220 nm to 1600 nm