Learn More
Fisherbrand™ UV Trays
Accessory for Fisherbrand Horizontal Gel Electrophoresis Systems.
Brand:  Fisherbrand™ MS7-UV10
 View more versions of this product
Product Code. 11837573
  • 470.00 NOK / Each
Only null left
Estimated Shipment: 18-07-2025
Add to basket
Description
Description
  • Accessory for Fisherbrand Horizontal Gel Electrophoresis Systems
Specifications
Specifications
Sub-Gel Midi Horizontal Gel Electrophoresis System
UV Tray
70mm x 100mm
Videos
  • Fisherbrand and Fisher Bioreagents - Focus on electrophoresis
    Fisherbrand provides a comprehensive range of horizontal gel systems for all your nucleic acid gel electrophoresis needs.
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
      Fisherbrand and Fisher BioReagents Focus on electrophoresis
    Certificates
    Promotions
    Promotions
    Product Content Correction
    Product Title
    Fisherbrand™ UV Trays > Dimensions (L x W): 70 x 100mm