Learn More
Fisherbrand™ UV Trays
Accessory for Fisherbrand Horizontal Gel Electrophoresis Systems.
Brand:  Fisherbrand™ MS20-UV25
 View more versions of this product
Product Code. 11833473
  • 1130.00 NOK / Each
Only null left
Estimated Shipment: 17-07-2025
Add to basket
Description
Description
  • Accessory for Fisherbrand Horizontal Gel Electrophoresis Systems
Specifications
Specifications
Sub-Gel Maxi Horizontal Gel Electrophoresis System
UV Tray
200mm x 250mm
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Fisherbrand™ UV Trays > Dimensions (L x W): 200 x 250mm