missing translation for 'onlineSavingsMsg'
Learn More

Biochrom UV Grade Silica Semi-micro Cuvette with Lid and Black Walls

UV Grade Silica Semi-micro Cuvette with Lid and Black Walls features UV grade silica construction.

Brand:  Biochrom 80-2109-82

 View more versions of this product

Product Code. 11745195

  • 66560.00 NOK / Pack of 8
Estimated Shipment: 13-11-2024
to see stock.
Add to basket
This item is not returnable. View return policy

Description

Description

  • UV Grade Silica construction
  • 10 mm semi-micro cell with lid and black walls
  • 10 mm pathlength
  • 4 mm internal width
  • For use with Ultrospec 7000/8000/9000, Ultrospec 2100 pro, Novaspec III/Plus, GeneQuant 1300/100
Specifications

Specifications

Matched Semi-Micro Standard Cuvette
UV Grade Silica
8 Pk.
190 to 1100 nm
Spectrophotometer, Photometer
Spectrophotometric Measurements
12.7 mm
400 μL
4 mm
10 mm
45 mm
12.5 mm
Rectangle
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers