missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP40 Monoclonal Antibody (OTI4F7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809863S
Additional Details : Weight : 0.05000kg
Description
USP40 Monoclonal antibody specifically detects USP40 in Human samples. It is validated for Western BlotSpecifications
USP40 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q96BW1 | |
Mouse | |
Affinity chromatography | |
RUO | |
139596 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Western Blot | |
OTI4F7 | |
Unconjugated | |
UPRT | |
B230215L03Rik; C730029K03; deubiquitinating enzyme 40; LRRGT00071; Ubiquitin carboxyl-terminal hydrolase 40; ubiquitin specific peptidase 40; ubiquitin specific protease 40; ubiquitin thioesterase 40; ubiquitin thiolesterase 40; ubiquitin-specific-processing protease 40; USP40 | |
Full length human recombinant protein of human UPRT produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |