missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP40 Monoclonal Antibody (OTI4F7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809863
Additional Details : Weight : 0.05000kg
Description
May be catalytically inactive.Specifications
USP40 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q9NVE5 | |
USP40 | |
Human recombit protein fragment corresponding to amino acids 767-912 of human USP40 produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot | |
OTI4F7 | |
Unconjugated | |
USP40 | |
ubiquitin specific peptidase 40 | |
Mouse | |
Affinity Chromatography | |
RUO | |
55230 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |