missing translation for 'onlineSavingsMsg'
Learn More

USP24 Rabbit anti-Human, Mouse, Rat, Clone: 7K7N2, Novus Biologicals™

Rabbit Monoclonal Antibody

1815.00 NOK - 4580.00 NOK

Specifications

Antigen USP24
Clone 7K7N2
Applications Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18339932
View Documents
Bio-Techne
NBP3-15286-20UL
20 μg
1815.00 NOK
20µL
Please to purchase this item. Need a web account? Register with us today!
18329194
View Documents
Bio-Techne
NBP3-15286-100UL
100 μg
4580.00 NOK
100µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

USP24 Monoclonal antibody specifically detects USP24 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications

Specifications

USP24
Western Blot
Unconjugated
Rabbit
Cell Biology
PBS, 0.05% BSA, 50% glycerol, pH7.3
23358
IgG
Affinity purified
7K7N2
Monoclonal
Purified
RUO
Human, Mouse, Rat
Deubiquitinating enzyme 24, EC 3.4.19.12, FLJ31309, KIAA1057ubiquitin carboxyl-terminal hydrolase 24, ubiquitin specific peptidase 24, ubiquitin specific protease 24, ubiquitin thioesterase 24, Ubiquitin thiolesterase 24, ubiquitin-specific processing protease 24, Ubiquitin-specific-processing protease 24
A synthetic peptide corresponding to a sequence within amino acids 2500-2600 of human USP24 (Q9UPU5).
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.