missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP11 Rabbit anti-Human, Mouse, Rat, Clone: 8I1K10, Novus Biologicals™
Rabbit Monoclonal Antibody
1815.00 NOK - 4580.00 NOK
Specifications
Antigen | USP11 |
---|---|
Clone | 8I1K10 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18379651
|
Bio-Techne
NBP3-15777-20UL |
20 μg |
1815.00 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18329733
|
Bio-Techne
NBP3-15777-100UL |
100 μg |
4580.00 NOK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
USP11 Monoclonal antibody specifically detects USP11 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
USP11 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cell Biology, Direct Reversal of DNA Damage | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
8237 | |
IgG | |
Affinity purified |
8I1K10 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
Deubiquitinating enzyme 11, EC 3.1.2.15, EC 3.4.19.12, ubiquitin carboxyl-terminal hydrolase 11, ubiquitin specific peptidase 11, ubiquitin specific protease 11, ubiquitin thioesterase 11, Ubiquitin thiolesterase 11, ubiquitin-specific processing protease 11, Ubiquitin-specific-processing protease 11, UHX1ubiquitin carboxyl-terminal hydrolase, X-linked | |
A synthetic peptide corresponding to a sequence within amino acids 800-900 of human USP11 (P51784). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title