missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ USF1 Recombinant Protein

Brand:  Abnova™ H00007391-P01.25ug

 View more versions of this product

Product Code. 16101792

  • 6245.00 NOK / 25µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH35505
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
60.2
8
Glutathione Sepharose 4 Fast Flow
25 μg
MKGQQKTAETEEGTVQIQEGAVATGEDPTSVAIASIQSAATFPDPNVKYVFRTENGGQVMYRVIQVSEGQLDGQTEGTGAISGYPATQSMTQAVIQGAFTSDDAVDTEGTAAETHYTYFPSTAVGDGAGGTTSGSTAAVVTTQGSEALLGQATPPGTGQFFVMMSPQEVLQGGSQRSIAPRTHPYSPKSEAPRTTRDEKRRAQHNEVERRRRDKINNWIVQLSKIIPDCSMESTKSGQSKGGILSKACDYIQELRQSNHRLSEELQGLDQLQLDNDVLRQQVEDLKNKNLLLRAQLRHHGLEVVIKNDSN
FCHL/FCHL1/HYPLIP1/MLTF/MLTFI/UEF/bHLHb11
USF1
Wheat Germ (in vitro)
GST
Antibody Production, Protein Array, ELISA, Western Blot
7391
USF1 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
USF1
Human
Recombinant
Solution
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers