Learn More
Memmert™ Usb User-Id Stick (With User-Id Licence), For Hcp50
Memmert™ USB User-ID Stick prevents undesired manipulation by unauthorized third parties.
Brand:  Memmert™ HCP50B33170
Product Code. 16344867
  • 3090.00 NOK / Each
Estimated Shipment: 05-08-2025
Explore more special offers
Add to basket
Includes: User-ID License
Description
Description
  • USB User-ID stick (with User-ID licence)
  • Oven-linked authorisation licence (User-ID-programme) on Memory-stick
  • Prevents undesired manipulation by unauthorised third parties
  • When reordering please specify serial number (only for units with TwinDISPLAY)
Specifications
Specifications
Only for units with TwinDISPLAY
Units With TwinDISPLAY
USB Stick
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Memmert™ Usb User-Id Stick (With User-Id Licence), For Hcp50 >