Learn More
Memmert™ USB-Ethernet Adapter for UN30
USB-Ethernet Adapter accessory for Memmert Ovens.
Brand: Memmert™ UN30E06192
This item is not returnable.
View return policy
Description
- Accessory
- For use with Memmert Ovens
Specifications
UN30 Oven | |
USB Ethernet Adapter |
Product Suggestions
Customers who viewed this item also viewed.
Product Content Correction
Product Title
Memmert™ USB-Ethernet Adapter for UN30 > For Use With: UN30