missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Uracil, 99+% Product Code.: 11483037

Thermo Scientific Chemicals Uracil, 99+%

Código de producto. 11483037
250 g, 250g
Click to view available options
Quantity:
50 g
250 g
1000 g
Tamaño de la unidad:
1000g
250g
50g
Este artículo no se puede devolver. Vea la política de devoluciones

Product Code. 11483037

Brand: Thermo Scientific Chemicals A15570.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

CAS: 66-22-8 | C4H4N2O2 | 112.09 g/mol

Uracil is produced by hydrolysis of nucleic acid and used in biochemical research. It serves as a allosteric regulator and coenzyme for reactions in the human body and in plants. It reacts with fluorine to get 5-fluorouracil, which is an anticancer drug. Further, it is used in drug delivery and in pharmaceuticals. In addition to this, it is used in the synthesis of caffeine and in the determination of microbial contamination of tomatoes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Uracil is produced by hydrolysis of nucleic acid and used in biochemical research. It serves as a allosteric regulator and coenzyme for reactions in the human body and in plants. It reacts with fluorine to get 5-fluorouracil, which is an anticancer drug. Further, it is used in drug delivery and in pharmaceuticals. In addition to this, it is used in the synthesis of caffeine and in the determination of microbial contamination of tomatoes.

Solubility
Soluble in water, aqueous ammonia, 1M sodium hydroxide solution and alkalies. Insoluble in ethanol and ether.

Notes
Incompatible with water, air and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 66-22-8
Molecular Formula C4H4N2O2
Molecular Weight (g/mol) 112.09
MDL Number MFCD00006016
InChI Key ISAKRJDGNUQOIC-UHFFFAOYSA-N
Synonym uracil, 2,4-dihydroxypyrimidine, 2,4 1h,3h-pyrimidinedione, 2,4-pyrimidinediol, 2,4-dioxopyrimidine, pyrimidine-2,4 1h,3h-dione, pirod, 2,4-pyrimidinedione, pyrod, hybar x
PubChem CID 1174
ChEBI CHEBI:17568
IUPAC Name 1H-pyrimidine-2,4-dione
SMILES O=C1NC=CC(=O)N1

Especificaciones

Melting Point ∼330°C
Assay Percent Range ≥99%
Quantity 250 g
Beilstein 606623
Merck Index 14,9850
Solubility Information Soluble in water,aqueous ammonia,1M sodium hydroxide solution and alkalies. Insoluble in ethanol and ether.
Formula Weight 112.09
Percent Purity ≥99%
Chemical Name or Material Uracil

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot Number10255037 Certificate TypeCertificate of Analysis Date17/05/2025 Product CodeA15570.30,A15570.0B,A15570.18
Lot Number10247276 Certificate TypeCertificate of Analysis Date17/07/2023 Product CodeA15570.30,A15570.0B,A15570.18
Lot Number10061714 Certificate TypeCertificate of Analysis Date23/11/2021 Product CodeA15570.30,A15570.0B,A15570.18
Lot Number10099116 Certificate TypeCertificate of Analysis Date23/11/2021 Product CodeA15570.30,A15570.0B,A15570.18
Lot Number10113898 Certificate TypeCertificate of Analysis Date23/11/2021 Product CodeA15570.30,A15570.0B,A15570.18

RUO – Research Use Only

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto
Thermo Scientific Chemicals Uracil, 99+% >

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado