Learn More
Fisherbrand™ Upright ULT Freezer Sliding Tray Racks
Product Code. 15769559 Shop All Fisherbrand Products
Click to view available options
Holds:
12 Boxes (2 in.)
15 (2in.) boxes
15 (3 in.) boxes
16 (2in.) boxes
20 (2in.) boxes
25 (2in.) boxes
28 (2in.) boxes
Dimensions (L x W x H):
419 x 140 x 225 mm
419 x 140 x 280 mm
559 x 140 x 225 mm
559 x 140 x 280 mm
559 x 140 x 391 mm
675 x 140 x 280 mm
679 x 140 x 225 mm
679 x 140 x 241 mm
Unit Size:
Each
Product Code. 15769559
Only null left
Add to basket
Add to basket
Fisherbrand inventory control racks are ideal for all cold storage application. They fit all standard types of upright ULT freezers.
  • Holds standard cryoboxes
  • Made of high quality stainless steel
  • Sliding trays provide more flexibility and easy access to the boxes
Specifications
Product Type Sliding Tray Rack
Holds 15 (2in.) boxes
Material Stainless Steel
For Use With (Application) Standard 2 in. Cryoboxes
For Use With (Equipment) Upright ULT Freezer
Dimensions (L x W x H) 419 x 140 x 280 mm