Learn More
Universol- ES liquid scintillation cocktail, MP Biomedicals™
Product Code. 15772252 Shop All MP Biomedicals Products
4 x 1 gal., 15.14 L
Click to view available options
Quantity:
1 gal.
4 x 1 gal.
Unit Size:
15.14 L
3.79 L
Specifications
Chemical Name or Material Universol- ES liquid scintillation cocktail
Density 0.9 to 1.0g/mL at 20°C
Quantity 4 x 1 gal.
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H360-May damage fertility or the unborn child.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P321-Specific treatment (see on this label).
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction
Product Title
Universol- ES liquid scintillation cocktail, MP Biomedicals™ >