Learn More
VELP Scientifica™ Universal H-Stand
The Universal H-Stand ensures stability and compatibility with all VELP™ Overhead Stirrers.
Brand:  VELP Scientifica™ A00000366
3029.57 NOK valid until 2025-07-11
Use promo code "25721" to get your promotional price.
Product Code. 16584764
  • 3245.00 NOK / Each
Quality Analysis and Quality Control Testing
Use promo code 24140 at checkout
View Available Promotions
Only null left
Estimated Shipment: 28-07-2025
Description
  • Great solution for using overhead stirrers on laboratory benches
  • Maximum stability thanks to H-shape of the base
  • Maximum resistance with premium materials such as aluminum and stainless steel, properly treated to resist chemicals and corrosion
  • Possible to adjust stand in order to adapt it to uneven surface
Specifications
Specifications
Stirrer Stand
462 x 430 mm
Certificates
Promotions
Promotions
Product Content Correction
Product Title
VELP Scientifica™ Universal H-Stand > Universal H-Stand; Dimensions (LxWxH): 462 x 430 mm; Type: Stirrer Stand