Learn More
ABEBA™ Uni6 White Safety Trainers S3
ABEBA™ Uni6 White Safety Trainers S3 are made of white tear and water resistant fabric with a slip resistant PU/TPU outsole.
1265.00 NOK
Specifications
Certifications/Compliance | CE EN ISO 20345:2012 S3 SRC ESD |
---|---|
Closure Type | Laces |
Color | White |
Eyelets Material | Steel |
Liner Material | Breathable silver ions lining |
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
16367981
|
ABEBA™
31629/35 |
35 |
1265.00 NOK
1 pair |
Estimated Shipment: 05-08-2025
Log in to see stock. |
||||||
16377981
|
ABEBA™
31629/36 |
36 |
1265.00 NOK
1 pair |
Estimated Shipment: 05-08-2025
Log in to see stock. |
||||||
16387981
|
ABEBA™
31629/37 |
37 |
1265.00 NOK
1 pair |
Estimated Shipment: 05-08-2025
Log in to see stock. |
||||||
16397981
|
ABEBA™
31629/38 |
38 |
1265.00 NOK
1 pair |
Estimated Shipment: 05-08-2025
Log in to see stock. |
||||||
16307991
|
ABEBA™
31629/39 |
39 |
1265.00 NOK
1 pair |
Estimated Shipment: 05-08-2025
Log in to see stock. |
||||||
16317991
|
ABEBA™
31629/40 |
40 |
1265.00 NOK
1 pair |
Estimated Shipment: 08-08-2025
Log in to see stock. |
||||||
16327991
|
ABEBA™
31629/41 |
41 |
1265.00 NOK
1 pair |
Estimated Shipment: 05-08-2025
Log in to see stock. |
||||||
16337991
|
ABEBA™
31629/42 |
42 |
1265.00 NOK
1 pair |
Estimated Shipment: 05-08-2025
Log in to see stock. |
||||||
16347991
|
ABEBA™
31629/43 |
43 |
1265.00 NOK
1 pair |
Estimated Shipment: 01-08-2025
Log in to see stock. |
||||||
16357991
|
ABEBA™
31629/44 |
44 |
1265.00 NOK
1 pair |
Estimated Shipment: 05-08-2025
Log in to see stock. |
||||||
16367991
|
ABEBA™
31629/45 |
45 |
1265.00 NOK
1 pair |
Estimated Shipment: 05-08-2025
Log in to see stock. |
||||||
16377991
|
ABEBA™
31629/46 |
46 |
1265.00 NOK
1 pair |
Estimated Shipment: 05-08-2025
Log in to see stock. |
||||||
16387991
|
ABEBA™
31629/47 |
47 |
1265.00 NOK
1 pair |
Estimated Shipment: 05-08-2025
Log in to see stock. |
||||||
16397991
|
ABEBA™
31629/48 |
48 |
1265.00 NOK
1 pair |
Estimated Shipment: 05-08-2025
Log in to see stock. |
||||||
Description
- Comfort, safety and style
- Comply to EN ISO 20345 : 2012, S3, SRC
- ESD dissipative according to DIN EN 61340
- Slip resistant PU/TPU outsole
- Tear and water resistant fabric
- Steel toe cap (200 joules)
- Textile antipuncture midsole
- Breathable and antibacterial lining with silver ions
- Sanitized treatment
- Removable insole Active comfort
Specifications
CE EN ISO 20345:2012 S3 SRC ESD | |
White | |
Breathable silver ions lining | |
Textile | |
Polyurethane, Thermoplastic Polyurethane | |
Steel | |
EN ISO 20345 : 2012; DIN EN 61340 | |
Yes |
Laces | |
Steel | |
Padded | |
Slip and abrasion resistant, ESD dissipative | |
Trainer | |
1 Pair | |
Water |
Product Title
ABEBA™ Uni6 White Safety Trainers S3